Assay Details
Gene Symbol: AJAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023635913.1 | XP_023491681.1 | 1610 | 110 | ||
XM_023635914.1 | XP_023491682.1 | 2185 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
