Assay Details
Gene Symbol: BEGAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628393.1 | XP_023484161.1 | 1389 | 83 | ||
XM_023628394.1 | XP_023484162.1 | 671 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
