Assay Details
Gene Symbol: ADRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614807.3 | XP_005614864.2 | 982 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005614807.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
