Assay Details
Gene Symbol: CACNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631697.1 | XP_023487465.1 | 1672 | 81 | ||
XM_023631698.1 | XP_023487466.1 | 1134 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
