Assay Details
Gene Symbol: TRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638625.1 | XP_023494393.1 | 1213 | 86 | ||
XM_023638626.1 | XP_023494394.1 | 1193 | 86 | |||
XM_023638627.1 | XP_023494395.1 | 1165 | 86 | |||
XM_023638628.1 | XP_023494396.1 | 1087 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
