Assay Details
Gene Symbol: NWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638241.1 | XP_023494009.1 | 6256 | 107 | ||
XM_023638242.1 | XP_023494010.1 | 8208 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
