Assay Details
Gene Symbol: KCNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641434.1 | XP_023497202.1 | 2416 | 94 | ||
XM_023641435.1 | XP_023497203.1 | 2362 | 94 | |||
XM_023641436.1 | XP_023497204.1 | 2838 | 94 | |||
XM_023641437.1 | XP_023497205.1 | 2475 | 94 | |||
XM_023641438.1 | XP_023497206.1 | 2554 | 94 | |||
XM_023641439.1 | XP_023497207.1 | 2361 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
