Assay Details
Gene Symbol: MALSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639364.1 | XP_023495132.1 | 459 | 76 | ||
XM_023639365.1 | XP_023495133.1 | 460 | 76 | |||
XM_023639367.1 | XP_023495135.1 | 463 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
