Assay Details
Gene Symbol: OPCML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001505111.6 | XP_001505161.1 | 1718 | 109 | ||
XM_023646454.1 | XP_023502222.1 | 1955 | 109 | |||
XM_023646455.1 | XP_023502223.1 | 1915 | 109 | |||
XM_023646456.1 | XP_023502224.1 | 2061 | 109 | |||
XM_023646457.1 | XP_023502225.1 | 1746 | 109 | |||
XM_023646459.1 | XP_023502227.1 | 1557 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
