Assay Details
Gene Symbol: MRPL40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613827.3 | XP_005613884.2 | 308 | 111 | ||
XM_023648101.1 | XP_023503869.1 | 308 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
