Assay Details
Gene Symbol: ATF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489102.6 | XP_001489152.1 | 374 | 85 | ||
XM_005609809.3 | XP_005609866.1 | 560 | 85 | |||
XM_005609810.3 | XP_005609867.1 | 441 | 85 | |||
XM_005609811.3 | XP_005609868.1 | 464 | 85 | |||
XM_023640750.1 | XP_023496518.1 | 514 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
