Assay Details
Gene Symbol: HSD11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490175.4 | XP_001490225.1 | 684 | 63 | ||
XM_014739666.2 | XP_014595152.1 | 680 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
