Assay Details
Gene Symbol: UCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014739655.2 | XP_014595141.1 | 5594 | 77 | ||
XM_023640801.1 | XP_023496569.1 | 852 | 77 | |||
XM_023640802.1 | XP_023496570.1 | 821 | 77 | |||
GenBank mRNA | JL629633.1 | - | 3 - 4 | 365 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
