Assay Details
Gene Symbol: MGST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493508.5 | XP_001493558.1 | 279 | 61 | ||
XM_014739654.2 | XP_014595140.1 | 314 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
