Assay Details
Gene Symbol: NECTIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503864.4 | XP_001503914.1 | 1144 | 79 | ||
XM_005609870.3 | XP_005609927.1 | 1146 | 79 | |||
XM_005609872.3 | XP_005609929.1 | 1145 | 79 | |||
XM_014739670.2 | XP_014595156.1 | 1003 | 79 | |||
XM_014739671.2 | XP_014595157.1 | 1001 | 79 | |||
XM_014739673.2 | XP_014595159.1 | 1005 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
