Assay Details
Gene Symbol: SLAMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504439.4 | XP_001504489.1 | 898 | 66 | ||
XM_005609956.3 | XP_005610013.1 | 904 | 66 | |||
XM_023640881.1 | XP_023496649.1 | 761 | 66 | |||
XM_023640882.1 | XP_023496650.1 | 769 | 66 | |||
XM_023640883.1 | XP_023496651.1 | 904 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
