Assay Details
Gene Symbol: KIRREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640929.1 | XP_023496697.1 | 1980 | 77 | ||
XM_023640930.1 | XP_023496698.1 | 1932 | 77 | |||
XM_023640931.1 | XP_023496699.1 | 1884 | 77 | |||
XM_023640932.1 | XP_023496700.1 | 1830 | 77 |
