Assay Details
Gene Symbol: KIRREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640929.1 | XP_023496697.1 | 2089 | 67 | ||
XM_023640930.1 | XP_023496698.1 | 2041 | 67 | |||
XM_023640931.1 | XP_023496699.1 | 1993 | 67 | |||
XM_023640932.1 | XP_023496700.1 | 1939 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
