Assay Details
Gene Symbol: CC2D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641813.1 | XP_023497581.1 | 4651 | 96 | ||
XM_023641820.1 | XP_023497588.1 | 4320 | 96 | |||
XM_023641830.1 | XP_023497598.1 | 4294 | 96 | |||
XM_023641834.1 | XP_023497602.1 | 4567 | 96 | |||
XM_023641837.1 | XP_023497605.1 | 4562 | 96 | |||
XM_023641842.1 | XP_023497610.1 | 4441 | 96 | |||
XM_023641847.1 | XP_023497615.1 | 4633 | 96 | |||
XM_023641852.1 | XP_023497620.1 | 4308 | 96 | |||
XM_023641856.1 | XP_023497624.1 | 4282 | 96 | |||
XM_023641863.1 | XP_023497631.1 | 4174 | 96 | |||
XM_023641874.1 | XP_023497642.1 | 3147 | 96 | |||
XM_023641877.1 | XP_023497645.1 | 2319 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
