Assay Details
Gene Symbol: MIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734174.2 | XP_014589660.1 | 283 | 89 | ||
XM_023624352.1 | XP_023480120.1 | 283 | 89 | |||
XM_023624357.1 | XP_023480125.1 | 285 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
