Assay Details
Gene Symbol: MBOAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650367.1 | XP_023506135.1 | 1165 | 93 | ||
XM_023650368.1 | XP_023506136.1 | 970 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
