Assay Details
Gene Symbol: BANF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654477.1 | XP_023510245.1 | 234 | 117 | ||
XM_023654478.1 | XP_023510246.1 | 274 | 117 | |||
XM_023654480.1 | XP_023510248.1 | 198 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
