Assay Details
Gene Symbol: TMEM80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598391.3 | XP_005598448.1 | 1641 | 62 | ||
XM_023654768.1 | XP_023510536.1 | 1983 | 62 | |||
XM_023654769.1 | XP_023510537.1 | 1071 | 62 | |||
XM_023654770.1 | XP_023510538.1 | 1058 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
