Assay Details
Gene Symbol: PDGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023655054.1 | XP_023510822.1 | 839 | 93 | ||
XM_023655055.1 | XP_023510823.1 | 781 | 93 | |||
XM_023655056.1 | XP_023510824.1 | 723 | 93 | |||
XM_023655057.1 | XP_023510825.1 | 723 | 93 | |||
XM_023655058.1 | XP_023510826.1 | 646 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
