Assay Details
Gene Symbol: HSD3B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598761.3 | XP_005598818.1 | 1121 | 68 | ||
XM_005598762.3 | XP_005598819.1 | 819 | 68 | |||
XM_014730005.2 | XP_014585491.1 | 1259 | 68 | |||
XM_014730006.2 | XP_014585492.1 | 957 | 68 | |||
XM_014730008.2 | XP_014585494.1 | 1135 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
