Assay Details
Gene Symbol: GEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619485.1 | XP_023475253.1 | 8319 | 113 | ||
XM_023619486.1 | XP_023475254.1 | 4567 | 113 | |||
XM_023619487.1 | XP_023475255.1 | 5632 | 113 | |||
XM_023619488.1 | XP_023475256.1 | 1846 | 113 | |||
XM_023619489.1 | XP_023475257.1 | 1880 | 113 | |||
XM_023619490.1 | XP_023475258.1 | 1749 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
