Assay Details
Gene Symbol: TRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619924.1 | XP_023475692.1 | 446 | 91 | ||
XM_023619925.1 | XP_023475693.1 | 485 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
