Assay Details
Gene Symbol: ACKR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600791.2 | XP_005600848.1 | 143 | 69 | ||
XM_023620647.1 | XP_023476415.1 | 218 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005600791.2, XM_023620647.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
