Assay Details
Gene Symbol: TAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023630379.1 | XP_023486147.1 | 1752 | 88 | ||
XM_023630384.1 | XP_023486152.1 | 913 | 88 | |||
XM_023630391.1 | XP_023486159.1 | 788 | 88 | |||
XM_023630394.1 | XP_023486162.1 | 1754 | 88 | |||
XM_023630396.1 | XP_023486164.1 | 503 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
