Assay Details
Gene Symbol: E2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504222.4 | XP_001504272.2 | 503 | 71 | ||
XM_023635519.1 | XP_023491287.1 | 506 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
