Assay Details
Gene Symbol: TRAM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636649.1 | XP_023492417.1 | 1292 | 80 | ||
XM_023636650.1 | XP_023492418.1 | 1292 | 80 | |||
XM_023636651.1 | XP_023492419.1 | 2663 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
