Assay Details
Gene Symbol: MED26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625374.1 | XP_023481142.1 | 325 | 80 | ||
XM_023625375.1 | XP_023481143.1 | 7730 | 80 | |||
XM_023625376.1 | XP_023481144.1 | 4846 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
