Assay Details
Gene Symbol: POLE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628692.1 | XP_023484460.1 | 615 | 90 | ||
GenBank mRNA | JL616258.1 | - | 3 - 4 | 275 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
