Assay Details
Gene Symbol: SALL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608257.3 | XP_005608314.2 | 4678 | 86 | ||
XM_023636966.1 | XP_023492734.1 | 3607 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
