Assay Details
Gene Symbol: HOPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638043.1 | XP_023493811.1 | 281 | 107 | ||
XM_023638044.1 | XP_023493812.1 | 538 | 107 | |||
XM_023638045.1 | XP_023493813.1 | 396 | 107 | |||
XM_023638046.1 | XP_023493814.1 | 336 | 107 | |||
XM_023638047.1 | XP_023493815.1 | 251 | 107 | |||
XM_023638048.1 | XP_023493816.1 | 249 | 107 | |||
XM_023638049.1 | XP_023493817.1 | 277 | 107 | |||
XM_023638050.1 | XP_023493818.1 | 176 | 107 | |||
XM_023638051.1 | XP_023493819.1 | 285 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
