Assay Details
Gene Symbol: ETV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500763.5 | XP_001500813.3 | 311 | 69 | ||
XM_005610008.3 | XP_005610065.2 | 328 | 69 |

More Information
Set Membership: |
![]() ![]() |
