Assay Details
Gene Symbol: ETV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500763.5 | XP_001500813.3 | 666 | 85 | ||
XM_005610008.3 | XP_005610065.2 | 683 | 85 | |||
XM_023640954.1 | XP_023496722.1 | 573 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
