Assay Details
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498650.4 | XP_001498700.3 | 1605 | 56 | ||
XM_005610048.3 | XP_005610105.1 | 1595 | 56 | |||
XM_014739902.1 | XP_014595388.1 | 1768 | 56 | |||
XM_023641051.1 | XP_023496819.1 | 1797 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
