Assay Details
Gene Symbol: CKS1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003365011.4 | XP_003365059.1 | 154 | 78 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
