Assay Details
Gene Symbol: PMVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497675.5 | XP_001497725.2 | 711 | 81 | ||
XM_014739914.2 | XP_014595400.1 | 943 | 81 | |||
XM_014739915.2 | XP_014595401.1 | 832 | 81 | |||
XM_014739916.2 | XP_014595402.1 | 853 | 81 | |||
XM_014739918.2 | XP_014595404.1 | 743 | 81 | |||
XM_023641074.1 | XP_023496842.1 | 811 | 81 | |||
XM_023641075.1 | XP_023496843.1 | 802 | 81 | |||
XM_023641076.1 | XP_023496844.1 | 644 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
