Assay Details
Gene Symbol: DYRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640454.1 | XP_023496222.1 | 376 | 64 | ||
XM_023640455.1 | XP_023496223.1 | 253 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
