Assay Details
Gene Symbol: SMUG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504557.4 | XP_001504607.1 | 402 | 67 | ||
XM_005611255.2 | XP_005611312.1 | 553 | 67 | |||
XM_023643714.1 | XP_023499482.1 | 1369 | 67 | |||
XM_023643715.1 | XP_023499483.1 | 1380 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
