Assay Details
Gene Symbol: RAD23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645292.1 | XP_023501060.1 | 1093 | 77 | ||
XM_023645293.1 | XP_023501061.1 | 1089 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
