Assay Details
Gene Symbol: DDIAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492152.4 | XP_001492202.1 | 623 | 95 | ||
XM_023645645.1 | XP_023501413.1 | 719 | 95 | |||
XM_023645646.1 | XP_023501414.1 | 594 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
