Assay Details
Gene Symbol: PCDH11X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023633699.1 | XP_023489467.1 | 1142 | 104 | ||
XM_023633700.1 | XP_023489468.1 | 1095 | 104 | |||
XM_023633701.1 | XP_023489469.1 | 1142 | 104 | |||
XM_023633702.1 | XP_023489470.1 | 1142 | 104 | |||
XM_023633703.1 | XP_023489471.1 | 1142 | 104 | |||
XM_023633704.1 | XP_023489472.1 | 930 | 104 | |||
XM_023633705.1 | XP_023489473.1 | 1142 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
