Assay Details
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014733367.2 | XP_014588853.2 | 404 | 62 | ||
XM_023636994.1 | XP_023492762.1 | 467 | 62 | |||
XM_023637001.1 | XP_023492769.1 | 404 | 62 | |||
XM_023637008.1 | XP_023492776.1 | 588 | 62 | |||
XM_023637011.1 | XP_023492779.1 | 508 | 62 | |||
XM_023637016.1 | XP_023492784.1 | 467 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
