Assay Details
Gene Symbol: BCCIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488987.4 | XP_001489037.1 | 620 | 55 | ||
XM_005602162.3 | XP_005602219.1 | 549 | 55 | |||
XM_023637244.1 | XP_023493012.1 | 534 | 55 | |||
XM_023637250.1 | XP_023493018.1 | 464 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
