Assay Details
Gene Symbol: ZRANB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489580.6 | XP_001489630.2 | 2089 | 63 | ||
XM_014733167.2 | XP_014588653.1 | 1353 | 63 | |||
XM_023637362.1 | XP_023493130.1 | 2373 | 63 | |||
XM_023637369.1 | XP_023493137.1 | 2242 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
