Assay Details
Gene Symbol: LHPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489690.4 | XP_001489740.2 | 519 | 67 | ||
XM_005602168.3 | XP_005602225.2 | 519 | 67 | |||
XM_023637462.1 | XP_023493230.1 | 524 | 67 | |||
XM_023637470.1 | XP_023493238.1 | 520 | 67 | |||
XM_023637480.1 | XP_023493248.1 | 520 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
