Assay Details
Gene Symbol: CHST15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916018.5 | XP_001916053.2 | 2153 | 81 | ||
XM_023637618.1 | XP_023493386.1 | 885 | 81 | |||
XM_023637628.1 | XP_023493396.1 | 2060 | 81 | |||
XM_023637635.1 | XP_023493403.1 | 1994 | 81 | |||
XM_023637643.1 | XP_023493411.1 | 1631 | 81 | |||
XM_023637658.1 | XP_023493426.1 | 885 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
